Iodopentafluorobenzene, 99%, stabilised over copper
Iodopentafluorobenzene, 99%, stabilised over copper
Iodopentafluorobenzene, 99%, stabilised over copper
Thermo Scientific Chemicals

Iodopentafluorobenzene, 99%, stabilised over copper

CAS: 827-15-6 | C6F5I | 293.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 312200010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS827-15-6
IUPAC Name1,2,3,4,5-pentafluoro-6-iodobenzene
Molecular FormulaC6F5I
InChI KeyOPYHNLNYCRZOGY-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(I)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only