Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack
Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack
Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack
Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack
Thermo Scientific™

Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack

Carry out pharmaceutical and biotech applications with these LDPE dispensing tips for dropper bottles, which feature precise drip control.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
312759-000140 μL
312758-000150 μL
Catalog number 312759-0001
Price (USD)
288.49
Online exclusive
326.03
Save 37.54
Case of 2000
Capacity (Metric):
40 μL
Price (USD)
288.49
Online exclusive
326.03
Save 37.54
Case of 2000

Carry out pharmaceutical and biotech applications with Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles. Tips feature precise drip control and can be used with Nalgene LDPE Dropper bottles and Colored PP Closures.

Features:
  • Low density polyethylene
  • Offered in two drop sizes (40μL or 50μL)
  • Fits Nalgene Dropper Bottles Series 312750 and 312751
  • Use with Nalgene dropper closures for color coding your reagents

Ordering Information:

Dropper Bottles Dispensing Tips are bulk packed 2000 per case. Order Nalgene dropper bottles (Cat. Nos. 312750, 312751) and colored closure (Cat. No. 312760) separately.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity (English)1.35 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle dispensing tips meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Diameter (Metric)11.22 mm
For Use With (Equipment)Nalgene Controlled Dropper
Height (English)0.65 in.
Height (Metric)16.5 mm
No. per Case2000
No. per Pack2000
Product TypeDropping Bottle Tips
Width (English)0.39 in.
Capacity (Metric)40 μL
Closure IncludedYes
ColorClear
Drop Size40/50 μL
MaterialLDPE
Unit SizeCase of 2000