Tris(pentafluorophenyl)phosphine, 97%
Tris(pentafluorophenyl)phosphine, 97%
Tris(pentafluorophenyl)phosphine, 97%
Thermo Scientific Chemicals

Tris(pentafluorophenyl)phosphine, 97%

CAS: 1259-35-4 | C18F15P | 532.15 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 313160010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1259-35-4
IUPAC Nametris(2,3,4,5,6-pentafluorophenyl)phosphane
Molecular FormulaC18F15P
InChI KeyFQLSDFNKTNBQLC-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
さらに表示
Infrared spectrumConforms
Appearance (Color)White to beige to light brown
Melting point108°C to 119°C
Appearance (Form)Powder or crystals
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only