DMEM/F-12, GlutaMAX™ Supplement, 500 mL - FAQs
DMEM/F-12, GlutaMAX™ Supplement, 500 mL - FAQs
View additional product information for DMEM/F-12, GlutaMAX™ supplement - FAQs (31331028)
3 product FAQs found
Do you offer DMEM/F12 medium without folic acid?
We do not offer DMEM/F12 medium without folic acid as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an “official” order or declining.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?
Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?
In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.