Isopropyldimethylchlorosilane, 95%
Isopropyldimethylchlorosilane, 95%
Isopropyldimethylchlorosilane, 95%
Thermo Scientific Chemicals

Isopropyldimethylchlorosilane, 95%

CAS: 3634-56-8 | C5H13ClSi | 136.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
31357025025 gGlass bottle
Catalog number 313570250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3634-56-8
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4150 to 1.4170 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Colorless
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only