Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap, 50mL - FAQs
Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap, 50mL - FAQs
View additional product information for Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap - FAQs (3139-0016, 3139-0030PK, 3139-0010, 3139-0016PK, 3139-0030, 3139-0050, 3139-0010PK, 3139-0050PK)
2 product FAQs found
Are there replacement O rings for Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap?
Replacement sealing-cap assemblies are available for selected sizes. We recommend Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0020) for 30 mL tubes and Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0024) for 50 mL tubes.
The actual brim capacity of the Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap is 42 mL, but it is designed to fit a standard rotor.
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.