1-Adamantyl isocyanate, 98%
1-Adamantyl isocyanate, 98%
1-Adamantyl isocyanate, 98%
Thermo Scientific Chemicals

1-Adamantyl isocyanate, 98%

CAS: 4411-25-0 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 314360250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4411-25-0
IUPAC Name1-isocyanatoadamantane
Molecular FormulaC11H15NO
InChI KeyVBHCPGFCIQDXGZ-UHFFFAOYSA-N
SMILESO=C=NC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point140°C to 150°C
Infrared spectrumConforms
Appearance (Color)White
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only