N-Propylethylenediamine, 99%
N-Propylethylenediamine, 99%
N-Propylethylenediamine, 99%
Thermo Scientific Chemicals

N-Propylethylenediamine, 99%

CAS: 111-39-7 | C5H14N2 | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 314750250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-39-7
IUPAC Name(2-aminoethyl)(propyl)amine
Molecular FormulaC5H14N2
InChI KeyCFNHVUGPXZUTRR-UHFFFAOYSA-N
SMILESCCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4394 to 1.4414 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only