Boric acid, 99.99%, (trace metal basis), extra pure
Boric acid, 99.99%, (trace metal basis), extra pure
Boric acid, 99.99%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Boric acid, 99.99%, (trace metal basis), extra pure

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 315185000
Price (USD)
188.65
Special offer
Online exclusive
Ends: 28-Dec-2026
222.00
Save 33.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
188.65
Special offer
Online exclusive
Ends: 28-Dec-2026
222.00
Save 33.35
Each
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=98.5 %
Appearance (Color)White
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acid
RUO – Research Use Only