Mercury(II) oxide, yellow, ACS reagent
Mercury(II) oxide, yellow, ACS reagent
Mercury(II) oxide, yellow, ACS reagent
Thermo Scientific Chemicals

Mercury(II) oxide, yellow, ACS reagent

CAS: 21908-53-2 | HgO | 216.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 316795000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS21908-53-2
IUPAC Nameoxomercury
Molecular FormulaHgO
InChI KeyUKWHYYKOEPRTIC-UHFFFAOYSA-N
SMILESO=[Hg]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=99.0 %
Iron (Fe)=<30 ppm
Appearance (Form)Powder
Nitrogen compounds (as N)=<0.005 %
Sulfate (SO4)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only