RPMI 1640 Medium, no glutamine
Gibco™

RPMI 1640 Medium, no glutamine

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as aRead more
Have Questions?
Catalog NumberQuantity
31870017100 mL
Catalog number 31870017
Price (USD)
17.03
Each
Quantity:
100 mL
Customize this product
Price (USD)
17.03
Each

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI 1640 is modified as follows:
WithWithout
• Phenol Red• L-glutamine
 • HEPES


The complete formulation is available.

Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12 and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeRPMI 1640
Quantity100 mL
Shelf Life12 months from date of manufacture
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesPhenol Red
Without AdditivesNo Glutamine, No HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.