2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, Thermo Scientific Chemicals
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, Thermo Scientific Chemicals
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
3196300505 gGlass bottle
Catalog number 319630050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19184-65-7
IUPAC Name2-(bromomethyl)-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC5H11BrO3
InChI KeyMMHHBAUIJVTLFZ-UHFFFAOYSA-N
SMILESOCC(CO)(CO)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to light beige
Melting point70°C to 77°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only