L(-)-Glutathione, oxidized, 98%
L(-)-Glutathione, oxidized, 98%
L(-)-Glutathione, oxidized, 98%
Thermo Scientific Chemicals

L(-)-Glutathione, oxidized, 98%

CAS: 27025-41-8 | C20H32N6O12S2 | 612.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number 320220050
Price (USD)
241.65
Online Exclusive
268.00
Save 26.35 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
241.65
Online Exclusive
268.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS27025-41-8
IUPAC Name(2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid
Molecular FormulaC20H32N6O12S2
InChI KeyYPZRWBKMTBYPTK-IOKZUGQQNA-N
SMILESN[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Loss on drying=<8 % (105°C, 3 h)
Appearance (Form)Powder
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only