Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals

Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D

CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3206500757.5 mLAmpule
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 320650075
Price (USD)
-
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Chemical Identifiers
CAS2206-26-0
IUPAC Name(²H₃)acetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES[2H]C([2H])([2H])C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.030 % (Coulometric)
D-Enrichment>=99.85 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only