Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals

Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D

CAS: 7789-20-0 | H2O | 20.03 g/mol
Have Questions?
Catalog NumberQuantity
3207100757.5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 320710075
Price (USD)
-
Quantity:
7.5 mL
Request bulk or custom format
Chemical Identifiers
CAS7789-20-0
IUPAC Name[(²H)oxy](²H)
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES[2H]O[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Tritium (T)=<74 Bq/g
Appearance (Color)Clear colorless
D-Enrichment>=99.85 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only