Di-tert-butylchlorophosphine, 96%
Di-tert-butylchlorophosphine, 96%
Di-tert-butylchlorophosphine, 96%
Thermo Scientific Chemicals

Di-tert-butylchlorophosphine, 96%

CAS: 13716-10-4 | C8H18ClP | 180.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 320901000
Price (EUR)
688,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
688,00
Each
Chemical Identifiers
CAS13716-10-4
IUPAC Namedi-tert-butyl(chloro)phosphane
Molecular FormulaC8H18ClP
InChI KeyMCRSZLVSRGTMIH-UHFFFAOYSA-N
SMILESCC(C)(C)P(Cl)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only