Tetravinyl tin, 95%
Tetravinyl tin, 95%
Tetravinyl tin, 95%
Thermo Scientific Chemicals

Tetravinyl tin, 95%

CAS: 1112-56-7 | C8H12Sn | 226.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 321080050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1112-56-7
IUPAC Nametetraethenylstannane
Molecular FormulaC8H12Sn
InChI KeyMZIYQMVHASXABC-UHFFFAOYSA-N
SMILESC=C[Sn](C=C)(C=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5000 to 1.5030 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only