Which should I use: the Pierce Western Blot Signal Enhancer or the Miser Antibody Extender Solution NC?
If you are happy with your western signal, you can use the Miser Antibody Extender Solution NC to minimize the amount of antibody you need to use and therefore reduce your costs. If you need to increase sensitivity, then you can use the Pierce Western Blot Signal Enhancer.