Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack, 8mL - FAQs
Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack, 8mL - FAQs
View additional product information for Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack - FAQs (322006-9125, 322006-9025, 322006-9050)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.