Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Thermo Scientific™

Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays

Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
322020-20002 L53B
322020-003030 mL20-415 mm
322020-006060 mL24-415 mm
322020-0125125 mL38-430 mm
322020-0250250 mL38-430 mm
322020-0500500 mL38-430 mm
322020-10001 L38-430 mm
Catalog number 322020-2000
Price (USD)
217.17
Online exclusive
248.40
Save 31.23
Case of 12
Capacity (Metric):
2 L
Closure Size:
53B
Price (USD)
217.17
Online exclusive
248.40
Save 31.23
Case of 12

Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.

  • Heavy-walled and durable
  • Shatter-resistant
  • Provide excellent gas barrier properties
  • In shrink-wrap trays
  • Molded in graduations
  • Clear bottles for easy identification of bottles
  • Nonsterile
  • Leakproof†

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity (English)67.6 oz.
Certifications/ComplianceUSP Class VI, FDA Compliance 21 CFR
Closure MaterialHDPE
Closure Size53B
Description2000mL, 53B closure, tray pack
Diameter (English)4.57 in.
Diameter (English) Inner Neck1.5 in.
Diameter (Metric)116 mm
Diameter (Metric) Inner Neck39 mm
For Use With (Application)Storing Media and Sera
Height (English)10.67 in.
Height (Metric)271 mm
No. per Case12
No. per Pack6
Product TypeMedia Bottle
ShapeSquare
Capacity (Metric)2 L
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPETG
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 12