Dipotassium hydrogen phosphate, 98.0-100.5% (dried basis),USP, Multi-Compendial, Endotoxin-free, GMP, J.T.Baker™
Avantor™

Dipotassium hydrogen phosphate, 98.0-100.5% (dried basis),USP, Multi-Compendial, Endotoxin-free, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
5 kg
12 kg
Número de catálogo 3250-07
Precio (CLP)
-
Cantidad:
12 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorBlanco
Punto de fusión340°C
EnvasePolyethylene pail, screw top
GradoMulticomponente/USP
Peso molecular (g/mol)174.18
Porcentaje de pureza98 to 100.5%
Forma físicaSolid
Cantidad12 kg
pH8.5 to 9.6
Unit SizeEach