trans,trans-2,4-Hexadien-1-ol, 99%, stabilized
trans,trans-2,4-Hexadien-1-ol, 99%, stabilized
trans,trans-2,4-Hexadien-1-ol, 99%, stabilized
Thermo Scientific Chemicals

trans,trans-2,4-Hexadien-1-ol, 99%, stabilized

CAS: 17102-64-6 | C6H10O | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 325570500
Price (USD)
214.65
Special offer
Online exclusive
Ends: 28-Dec-2026
252.00
Save 37.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
214.65
Special offer
Online exclusive
Ends: 28-Dec-2026
252.00
Save 37.35
Each
Chemical Identifiers
CAS17102-64-6
IUPAC Name(2E,4E)-hexa-2,4-dien-1-ol
Molecular FormulaC6H10O
InChI KeyMEIRRNXMZYDVDW-MQQKCMAXSA-N
SMILESC\C=C\C=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer0.1 % alpha-tocopherol
Appearance (Color)Clear colorless to white to light yellow
Appearance (Form)Low melting solid or liquid after melting
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only