Dichloromethane, for residue and pestic. anal., stab. with amylene
Dichloromethane, for residue and pestic. anal., stab. with amylene
Dichloromethane, for residue and pestic. anal., stab. with amylene
Dichloromethane, for residue and pestic. anal., stab. with amylene
Thermo Scientific Chemicals

Dichloromethane, for residue and pestic. anal., stab. with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
4 L
Catalog number 326600010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 % (K.F.)
GC(ECD) Passes test
StabilizerApprox. 50 ppm amylene
Appearance (Color)Clear colorless
Residue after evaporation=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only