Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals

Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326820010
Price (JPY)
-
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 %
Stabilizer50 to 150 ppm Amylene
Appearance (Color)Clear colorless
GC>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only