Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
327081000100 gPlastic bottle
Catalog number 327081000
Price (USD)
61.80
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
61.80
Each
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Iodometric>=99 %
Heavy metals (as Pb)=<50 ppm
Acidity=<0.2 % (H2SO4)
Dnase,Rnase,Protease activitynone detected
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only