Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Acros Organics

Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
Catalog number 327272500
also known as 32727
HSN 29051220
Price (INR)
8,230.00
Each
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
8,230.00
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.5 %
Water=<0.05 % (Coulometric)
Heavy metals (as Pb)=<1 ppm
Copper (Cu)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only