Diisopropyl azodicarboxylate, 94%
Diisopropyl azodicarboxylate, 94%
Diisopropyl azodicarboxylate, 94%
Thermo Scientific Chemicals

Diisopropyl azodicarboxylate, 94%

CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 327560050
Price (USD)
39.65
Online exclusive
44.00
Save 4.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online exclusive
44.00
Save 4.35
Each
Chemical Identifiers
CAS2446-83-5
IUPAC NameN-{[(propan-2-yloxy)carbonyl]imino}(propan-2-yloxy)formamide
Molecular FormulaC8H14N2O4
InChI KeyVVWRJUBEIPHGQF-UHFFFAOYSA-N
SMILESCC(C)OC(=O)N=NC(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4180 to 1.4220 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=92 %
Appearance (Color)Clear to hazy yellow to orange to red
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only