ACES, 99+%, for biochemistry
ACES, 99+%, for biochemistry
ACES, 99+%, for biochemistry
Thermo Scientific Chemicals

ACES, 99+%, for biochemistry

ACES, >-99%, C4H10N2O4S, CAS Number-7365-82-4, N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine, 1kg, 0.02 max. at 280nm, 0.04 max. at 260nm, (5% aq. soln.), (5% aq. soln.) at 0nm, 99+%, 100.0, 99.0, CHEBI:39062, 100ppm max., White, 230-908-4, 182.19, Biochemical | CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 327670010
Price (USD)
888.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,045.00
Save 156.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
888.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,045.00
Save 156.35
Each
Chemical Identifiers
CAS7365-82-4
IUPAC Name2-[(carbamoylmethyl)amino]ethane-1-sulfonic acid
Molecular FormulaC4H10N2O4S
InChI KeyDBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILESNC(=O)CNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.04
UVat 280 nm A: =<0.02
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for biochemistry
RUO – Research Use Only