This product ships via ground from US northeast.
MES monohydrate, 99+%, for biochemistry
MES monohydrate, 99+%, for biochemistry
MES monohydrate, 99+%, for biochemistry
Thermo Scientific Chemicals

MES monohydrate, 99+%, for biochemistry

CAS: 145224-94-8 | C6H15NO5S | 195.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 327761000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS145224-94-8
IUPAC Name4-(2-sulfonatoethyl)morpholin-4-ium
Molecular FormulaC6H13NO4S
InChI KeySXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CC[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Titration with NaOH99.0 to 101.0 %
Water8.0 to 9.0 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only