Rottlerin, 95+%
Rottlerin, 95+%
Rottlerin, 95+%
Thermo Scientific Chemicals

Rottlerin, 95+%

CAS: 82-08-6 | C30H28O8 | 516.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
32849010010 mgGlass bottle
Catalog number 328490100
Price (USD)
-
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS82-08-6
IUPAC Name(2E)-1-{6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethyl-2H-chromen-8-yl}-3-phenylprop-2-en-1-one
Molecular FormulaC30H28O8
InChI KeyDEZFNHCVIZBHBI-ZHACJKMWSA-N
SMILESCC(=O)C1=C(O)C(C)=C(O)C(CC2=C(O)C3=C(OC(C)(C)C=C3)C(C(=O)\C=C\C3=CC=CC=C3)=C2O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Orange to brown
Infrared spectrumConforms
HPLC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only