Glipizide, 98+%
Glipizide, 98+%
Glipizide, 98+%
Thermo Scientific Chemicals

Glipizide, 98+%

CAS: 29094-61-9 | C21H27N5O4S | 445.5 g/mol
Have Questions?
Catalog NumberQuantity
3291100101 g
Catalog number 329110010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS29094-61-9
IUPAC NameN-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-5-methylpyrazine-2-carboxamide
Molecular FormulaC21H27N5O4S
InChI KeyZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILESCC1=CN=C(C=N1)C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying=<1 % (100°C, 3 hrs) (vacuum)
Heavy metals=<50 ppm
HPLC>=98.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only