N-Methyl-D-aspartic acid, 98%
N-Methyl-D-aspartic acid, 98%
N-Methyl-D-aspartic acid, 98%
Thermo Scientific Chemicals

N-Methyl-D-aspartic acid, 98%

CAS: 6384-92-5 | C5H9NO4 | 147.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Catalog number 329192500
Price (EUR)
236,00
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
236,00
Each
Chemical Identifiers
CAS6384-92-5
IUPAC Name(2R)-2-(methylazaniumyl)butanedioate
Molecular FormulaC5H8NO4
InChI KeyHOKKHZGPKSLGJE-GSVOUGTGSA-M
SMILESC[NH2+][C@H](CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White to off-white
Specific optical rotation-26° to -30° (20°C, 589 nm) (c=1, 6 N HCl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only