5-chloro-2-nitrobenzaldehyde, 97%
5-chloro-2-nitrobenzaldehyde, 97%
5-chloro-2-nitrobenzaldehyde, 97%
Thermo Scientific Chemicals

5-chloro-2-nitrobenzaldehyde, 97%

CAS: 6628-86-0 | C7H4ClNO3 | 185.563 g/mol
Have Questions?
Catalog NumberQuantity
3297600505 g
Catalog number 329760050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6628-86-0
IUPAC Name5-chloro-2-nitrobenzaldehyde
Molecular FormulaC7H4ClNO3
InChI KeySWGPIDCNYAYXMJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point72°C to 80°C
Appearance (Form)Powder
Appearance (Color)Yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only