1-Hexyne, 98%
1-Hexyne, 98%
1-Hexyne, 98%
Thermo Scientific Chemicals

1-Hexyne, 98%

CAS: 693-02-7 | C6H10 | 82.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 329900250
Price (USD)
51.65
Online exclusive
57.40
Save 5.75
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Online exclusive
57.40
Save 5.75
Each
Chemical Identifiers
CAS693-02-7
IUPAC Namehex-1-yne
Molecular FormulaC6H10
InChI KeyCGHIBGNXEGJPQZ-UHFFFAOYSA-N
SMILESCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.3980 to 1.4000 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only