6,7-Dihydro-4(5H)-benzofuranone, 98%
6,7-Dihydro-4(5H)-benzofuranone, 98%
6,7-Dihydro-4(5H)-benzofuranone, 98%
Thermo Scientific Chemicals

6,7-Dihydro-4(5H)-benzofuranone, 98%

CAS: 16806-93-2 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 330080050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16806-93-2
IUPAC Name4,5,6,7-tetrahydro-1-benzofuran-4-one
Molecular FormulaC8H8O2
InChI KeyDXWQOYPYNPSVRL-UHFFFAOYSA-N
SMILESO=C1CCCC2=C1C=CO2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
GC>=97.5 %
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only