2-chloromalonaldehyde, 95%
2-chloromalonaldehyde, 95%
2-chloromalonaldehyde, 95%
Thermo Scientific Chemicals

2-chloromalonaldehyde, 95%

CAS: 36437-19-1 | C3H3ClO2 | 106.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 330120100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36437-19-1
IUPAC Name(2Z)-2-chloro-3-hydroxyprop-2-enal
Molecular FormulaC3H3ClO2
InChI KeySYXFXUMRQSTIKC-IWQZZHSRSA-N
SMILESOC=C(/Cl)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with TBAH>=94 %
Appearance (Color)Brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only