3,5-Dibromopyridine, 98+%
3,5-Dibromopyridine, 98+%
3,5-Dibromopyridine, 98+%
Thermo Scientific Chemicals

3,5-Dibromopyridine, 98+%

CAS: 625-92-3 | C5H3Br2N | 236.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 330131000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS625-92-3
IUPAC Name3,5-dibromopyridine
Molecular FormulaC5H3Br2N
InChI KeySOSPMXMEOFGPIM-UHFFFAOYSA-N
SMILESBrC1=CC(Br)=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Powder or crystals or needles
Melting point110°C to 114°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only