Search
View additional product information for Sulfo-SBED Biotin Label Transfer Kit - Western Blot Application - FAQs (33073)
1 product FAQs found
Currently, we offer sulfo-SBED Biotin Label Transfer Reagent (Cat. Nos. 33033 and 33034) as well as the Sulfo-SBED Biotin Label Transfer Kit for Western Blot Application (Cat. No. 33073).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.