2,6-Diacetylpyridine, 99%
2,6-Diacetylpyridine, 99%
2,6-Diacetylpyridine, 99%
Thermo Scientific Chemicals

2,6-Diacetylpyridine, 99%

CAS: 1129-30-2 | C9H9NO2 | 163.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 330860250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1129-30-2
IUPAC Name1-(6-acetylpyridin-2-yl)ethan-1-one
Molecular FormulaC9H9NO2
InChI KeyBEZVGIHGZPLGBL-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(=N1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point78°C to 82°C
Appearance (Form)Powder or crystals or needles
GC>=98.5 %
Appearance (Color)White to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only