4-Chloro-o-phenylenediamine, 97%
4-Chloro-o-phenylenediamine, 97%
4-Chloro-o-phenylenediamine, 97%
Thermo Scientific Chemicals

4-Chloro-o-phenylenediamine, 97%

CAS: 95-83-0 | C6H7ClN2 | 142.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 331861000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-83-0
IUPAC Name4-chlorobenzene-1,2-diamine
Molecular FormulaC6H7ClN2
InChI KeyBXIXXXYDDJVHDL-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Powder or granules and/or chunks
Melting point70°C to 75°C
Appearance (Color)Grey-brown to dark brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only