2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
Thermo Scientific Chemicals

2-Mercaptobenzothiazole, 97%

CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 332050250
Price (USD)
23.65
Online exclusive
25.70
Save 2.05
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
23.65
Online exclusive
25.70
Save 2.05
Each
Chemical Identifiers
CAS149-30-4
IUPAC Name2,3-dihydro-1,3-benzothiazole-2-thione
Molecular FormulaC7H5NS2
InChI KeyYXIWHUQXZSMYRE-UHFFFAOYSA-N
SMILESS=C1NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point177°C to 182°C
Solubilityyellow solution
Appearance (Color)White to yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only