Diethyl (2-aminoethyl)phosphonate oxalate, 97%
Diethyl (2-aminoethyl)phosphonate oxalate, 97%
Diethyl (2-aminoethyl)phosphonate oxalate, 97%
Thermo Scientific Chemicals

Diethyl (2-aminoethyl)phosphonate oxalate, 97%

CAS: 203192-98-7 | C6H17NO3P | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 332220050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS203192-98-7
IUPAC Namediethyl (2-azaniumylethyl)phosphonate
Molecular FormulaC6H17NO3P
InChI KeyJUAGHBASZWRMQH-UHFFFAOYSA-O
SMILESCCOP(=O)(CC[NH3+])OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Melting point135°C to 141°C
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only