CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System
CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System
CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System
Thermo Scientific™
CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System
CultiMaxx™ Shelving System to support Nunc™ Cell Factory™ systems
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Shelves
332236G015
332354G0110
332355G016
332239G013
501662685 (Reinforced)
501662853 (Reinforced)
Catalog number 332236G01
Price (CLP)
-
No. of Shelves:
5
These shelving systems are designed to support scale-out of the Nunc™ Cell Factory™ System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.
These shelving systems are designed to support scale-out of the Nunc™ Cell Factory™ System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5 mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.
Easy Installation
  • Shelving systems are retrofittable and can be self-installed.
Specifications
DescriptionShelving system to support Nunc™ Cell Factory™ with 4 layers for Forma™ Steri-Cult™ 3310/3311 323L CO2 Incubator
For Use With (Equipment)Forma Steri-Cult 3310/3311 CO2 Incubator
Includes5 shelves
TypeShelving System
MaterialStainless Steel
No. of Shelves5
Unit SizeEach