Nunc™ Transport Containers
Actual product may vary
Thermo Scientific™
Nunc™ Transport Containers
Transport 10-11mL centrifuge tubes safely with Thermo Scientific™ Nunc™ Transport Containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
333511Separate cap
333510Centrifuge Tube with Stopper
362820Foam insert
Catalog number 333511
Price (MXN)
-
Includes:
Separate cap
Transport 10-11mL centrifuge tubes safely with Thermo Scientific™ Nunc™ Transport Containers. Nunc Labware Products are made from high purity resins, and molded using our state of the art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
  • Containers made of high-impact polystyrene, polyethylene or polypropylene
  • Caps made of polyethylene
Specifications
Capacity (Metric)10 mL to 11 mL
Closure MaterialPolyethylene
DescriptionTransparent polypropylene container
For Use With (Application)Transporting 10 to 11mL centrifuge tubes
IncludesSeparate cap
Closure IncludedYes
MaterialPolypropylene
Product TypeTransport Container
Unit SizeEach