2-Hydroxyethyl acrylate, 97%, stabilized
2-Hydroxyethyl acrylate, 97%, stabilized
2-Hydroxyethyl acrylate, 97%, stabilized
Thermo Scientific Chemicals

2-Hydroxyethyl acrylate, 97%, stabilized

CAS: 818-61-1 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 334080025
Price (USD)
286.65
Online exclusive
318.00
Save 31.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
286.65
Online exclusive
318.00
Save 31.35
Each
Chemical Identifiers
CAS818-61-1
IUPAC Name2-hydroxyethyl prop-2-enoate
Molecular FormulaC5H8O3
InChI KeyOMIGHNLMNHATMP-UHFFFAOYSA-N
SMILESOCCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer200 to 450 ppm MEHQ
Appearance (Form)Clear liquid
Color scale=<30 APHA
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only