Polysorbate 40
Polysorbate 40
Polysorbate 40
Thermo Scientific Chemicals

Polysorbate 40

CAS: 9005-66-7 | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | 522.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 334142500
Price (TWD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-66-7
Molecular Formula(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
InChI KeyDVRFSZFBSOKHQW-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00165345
SpecificationsSpecification SheetSpecification Sheet
Hydroxyl value89 to 105 mg KOH/g
Appearance (Color)Clear to hazy light yellow to dark yellow
Infrared spectrumConforms
Appearance (Form)Viscous liquid or paste
Acid value=<3 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only