2-Naphthalenethiol, 98+%
2-Naphthalenethiol, 98+%
2-Naphthalenethiol, 98+%
Thermo Scientific Chemicals

2-Naphthalenethiol, 98+%

CAS: 91-60-1 | C10H8S | 160.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 334520050
Price (USD)
49.65
Online exclusive
54.50
Save 4.85
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Online exclusive
54.50
Save 4.85
Each
Chemical Identifiers
CAS91-60-1
IUPAC Namenaphthalene-2-thiol
Molecular FormulaC10H8S
InChI KeyRFCQDOVPMUSZMN-UHFFFAOYSA-N
SMILESSC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or flakes
Melting point79°C to 83°C
Appearance (Color)White to light beige to light grey
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only