4-Isopropylbenzaldehyde, 98%
4-Isopropylbenzaldehyde, 98%
4-Isopropylbenzaldehyde, 98%
Thermo Scientific Chemicals

4-Isopropylbenzaldehyde, 98%

CAS: 122-03-2 | C10H12O | 148.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 334885000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-03-2
IUPAC Name4-(propan-2-yl)benzaldehyde
Molecular FormulaC10H12O
InChI KeyWTWBUQJHJGUZCY-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Density(20°C) 0.975 to 0.983 g/ml
Infrared spectrumConforms
Refractive index1.5290 to 1.5320 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only