3-Bromocyclohexene, 95%
3-Bromocyclohexene, 95%
3-Bromocyclohexene, 95%
Thermo Scientific Chemicals

3-Bromocyclohexene, 95%

CAS: 1521-51-3 | C6H9Br | 161.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 335120100
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.10
Save 12.45
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.10
Save 12.45
Each
Chemical Identifiers
CAS1521-51-3
IUPAC Name3-bromocyclohex-1-ene
Molecular FormulaC6H9Br
InChI KeyAJKDUJRRWLQXHM-UHFFFAOYNA-N
SMILESBrC1CCCC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
Refractive index1.5280 to 1.5320 (20°C, 589 nm)
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only