Lithium azide, pure, 20% solution in water
Lithium azide, pure, 20% solution in water
Lithium azide, pure, 20% solution in water
Thermo Scientific Chemicals

Lithium azide, pure, 20% solution in water

CAS: 19597-69-4 | LiN3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 335360250
Price (USD)
137.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
137.00
Each
Chemical Identifiers
CAS19597-69-4
IUPAC Namelithium(1+) azide
Molecular FormulaLiN3
InChI KeyGUWHRJQTTVADPB-UHFFFAOYSA-N
SMILES[Li+].[N-]=[N+]=[N-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Titration Argentometric17 to 23 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only